Learn More
CD8 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: RPA-T8, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566452
 View more versions of this product
Catalog No. BDB566452
Add to cart
Description
Description
CD8 Monoclonal antibody specifically detects CD8 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD8
RPA-T8
Purified
IgG1, κ
RUO
Primary
Human, Baboon, Cynomolgus, Rhesus Macaque
Flow Cytometry
BB700
Mouse
100 Tests
Refrigerate (2 to 8°C)
Monoclonal