Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Mouse anti-Human, Pacific Orange™, Clone: 3B5, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MHCD0830

 View more versions of this product

Catalog No. 50-112-1204


Add to cart

Description

Description

PerCP conjugates are excited by the 488 nm line of the argon-ion laser and have a large Stokes shift, providing minimal spectral overlap into the R-PE (phycoerythrin) channel. This antibody recognizes the alpha chain alone as well as the alpha/beta heterodimer.

CD8 (Cluster of Differentiation 8) is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediate efficient cell-cell interactions within the immune system. The CD8 antigen acts as a co-receptor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20-35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer (NK) cells, subpopulations of peripheral blood null cells, thymocytes and bone marrow cells. The CD8 co-receptor functions as either a homodimer composed of two alpha chains, or as a disulfide-linked heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. The majority of CD8+ T cells express CD8 as a alpha/beta heterodimer. In HIV, the HIV-2 envelope glycoprotein binds CD8 alpha chain (but not the beta chain).
Specifications

Specifications

CD8
Monoclonal
Pacific Orange™
CD8B
Liquid
CD8A, CD8B
Human CD8
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
3B5
PBS with BSA and 0.1% sodium azide
P01732, P10966
CD8, Leu-2, MAL, p32, CD8a, CD8b, CD8alpha, CD8beta
Mouse
IgG2a
0.5 mL
4° C, store in dark
Primary
925, 926
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit