Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (2ST8.5H7), Novus Biologicals™
Mouse Monoclonal Antibody
$204.00 - $482.50
Specifications
Antigen | CD8 |
---|---|
Clone | 2ST8.5H7 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry Neat-1:10, Immunoprecipitation 1:10-1:500 |
Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017757
|
Novus Biologicals
NB100659280.025MG |
0.025 mg |
Each for $204.00
|
|
|||||
NB10065928
|
Novus Biologicals
NB10065928 |
0.1 mg |
Each for $482.50
|
|
|||||
Description
CD8 Monoclonal specifically detects CD8 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.Specifications
CD8 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Phospho Specific, Signal Transduction | |
P10966 | |
925 | |
Normal human T lymphocytes | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
2ST8.5H7 | |
Western Blot 1:100-1:2000, Flow Cytometry Neat-1:10, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Primate | |
CD8, CD8a molecule, LEU2, p32 | |
CD8A | |
IgG2a | |
Protein A or G purified | |
NB100-65928 recognizes the human CD8 cell surface antigen. The antibody binds to the beta chain, recognizing an epitope that is dependent upon expression of both CD8 alpha and CD8 beta. CD8 molecules are composed of dimers of either alpha/alpha or alpha/beta polypeptide combinations. Both types of dimer may co-exist at the cell surface. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title