Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Mouse anti-Rat, Alexa Fluor™ 350, Clone: OX-8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212523AF350

Catalog No. NP21223A30


Add to cart

Description

Description

CD8 Monoclonal antibody specifically detects CD8 in Rat samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications

Specifications

CD8
OX-8
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
CD8, CD8a molecule, LEU2, p32
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody OX-8 recognizes the hinge-like membrane-proximal domain of rat CD8a (32-34 kDa; alpha chain of the CD8 antigen).
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CD8A
High Mw glycoproteins from rat thymocytes
0.1 mL
Primary
925
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit