Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Rabbit anti-Human, Allophycocyanin/Cy7, Clone: C8/1779R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254595APCCY7

Catalog No. N254595APC7


Add to cart

Description

Description

CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD8
C8/1779R
Flow Cytometry
Conjugated, Purified
CD8A
Recombinant full-length human CD8a protein
Protein A purified
Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary
925.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
CD8, CD8a molecule, LEU2, p32
Rabbit
IgG Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit