Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Rabbit anti-Mouse, Alexa Fluor™ 750, Clone: YTS 105.18, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252659AF750

Catalog No. NP2269A70


Add to cart

Description

Description

CD8 Monoclonal antibody specifically detects CD8 in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen),Immunofluorescence.
Specifications

Specifications

CD8
YTS 105.18
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence
CD8, CD8a molecule, LEU2, p32
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
Recognizes a non polymorphic epitope on the mouse CD8 alpha chain. This antibody has been reported to block MHC I dependent T cell responses in vitro and in vivo.
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CD8A
Ly-2-transfected mouse L cells.
0.1 mL
Primary
925
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit