Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CD8, Rabbit Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD8, Rabbit Monoclonal Antibody.

$556.50 - $1252.00

Specifications

Antigen CD8
Clone SP16
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Rabbit
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
RM9116R7
View Documents
Thermo Scientific™
RM9116R7
7mL Each for $586.00
Add to cart
 
RM9116RQ
View Documents
Thermo Scientific™
RM9116RQ
12mL Each for $556.50
Add to cart
 
RM9116S
View Documents
Thermo Scientific™
RM9116S
1mL Each for $1,252.00
Add to cart
 
RM9116S1
View Documents
Thermo Scientific™
RM9116S1
500μL Each for $786.00
Add to cart
 
RM9116S0
View Documents
Thermo Scientific™
RM9116S0
100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
RM9116R1
View Documents
Thermo Scientific™
RM9116R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD8, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

 

CD8 molecule consists of two chains, termed α and β chain, which are expressed as a disulphide-linked α/β heterdimer or as an α/α homodimer on T cell subset, thymocytes and NK cells. The majority of CD8+ T cells express CD8 as α/β heterdimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 alpha chain (but not β chain).

Host Species: Rabbit

Clone: SP16

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of α-chain of the CD8 molecule

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD8
Immunohistochemistry (Paraffin)
Rabbit
T-Cell
Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of alpha chain of the CD8 molecule
SP16
Monoclonal
IVD
Primary
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit