Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CD8, Rabbit Polyclonal Antibody

Ensure accurate and reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD8, Rabbit Polyclonal Antibody.

Manufacturer:  Thermo Scientific™ RB9009P1

 View more versions of this product

Catalog No. RB9009P1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ensure accurate and reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD8, Rabbit Polyclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

CD8 molecule consists of two chains, termed alpha and Beta chain, which are expressed as a disulphide-linked alpha/Beta heterdimer or as an alpha/alpha homodimer on T cell subset, thymocytes and NK cells. The majority of CD8+ T cells express CD8 as alpha/Beta heterdimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 alpha chain (but not Beta chain).

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of α-chain of the CD8 molecule

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD8
Polyclonal
Unconjugated
Rabbit
IgG
IVD
Primary
Polyclonal
Immunohistochemistry (Paraffin)
200µg/mL
Concentrated
Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of alpha chain of the CD8 molecule
500μL
T-Cell
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit