Learn More
CD8 Rat anti-Mouse, Alexa Fluor™ 350, Clone: AP-MAB0708, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212183AF350
Catalog No. NP212183A30
Add to cart
Description
Description
CD8 Monoclonal antibody specifically detects CD8 in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
CD8
AP-MAB0708
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
CD8, CD8a molecule, LEU2, p32
Rat
IgG2b
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CD8A
This antibody was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse Thymus/Spleen Tissues.
0.1 mL
Primary
925