Learn More
CD80 Mouse anti-Human, PE, Clone: MEM-233, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD8004
Catalog No. 50-112-1460
Add to cart
Description
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28 and of an inhibitory receptor CTLA-4 (CD152). The both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, the both molecules can also substitute for each other in this process. The question what are the differences in CD80 and CD86 competency has not been fully elucidated yet; there are still conflicts in results about their respective roles in initiation or sustaining of the T cell immune response.
Specifications
Specifications
CD80
Monoclonal
PE
CD80
Liquid
CD80
Human CD80
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
MEM-233
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide
P33681
B7-1, B7.1, Cd28l, Ly-53, Ly53, MIC17, TS/A-1, LAB7, B7.1, CD28LG, CD28LG1
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
941