Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CD81 Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$285.00 - $1004.00

Specifications

Formulation PBS (pH 7.4) containing 10% glycerol. containingno Preservative
Gene Alias CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
Gene ID (Entrez) 975
Gene Symbol CD81
Name Recombinant CD81 Protein
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB60215250U View Documents Novus Biologicals™
NBP260215250UG
250μg Each for $1,004.00
Add to cart
 
NB6021550UG View Documents Novus Biologicals™
NBP26021550UG
50μg Each for $285.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 113-201 of Human CD81 The Recombinant Human CD81 Protein is derived from Baculovirus. The Recombinant Human CD81 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

PBS (pH 7.4) containing 10% glycerol. containingno Preservative
975
Recombinant CD81 Protein
Baculovirus
0.5mg/mL
SDS-Page
CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
CD81
Recombinant Protein
RUO
< 1.0 EU per 1ug of protein (determined by LAL method)

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit