Learn More
Abnova™ CD83 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00009308T01
Catalog No. 89-938-780
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CD83 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 22.66kDa
Transient overexpression cell lysate was tested with anti-CD83 antibody (Mfr. No. H00009308-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_004233.3
pCMV-CD83 full-length
100 μL
Human