Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD83 Antibody (MM0179-4B34), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212180JF646
Description
CD83 Monoclonal specifically detects CD83 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD83 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD83 | |
Human recombinant CD83 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
MM0179-4B34 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83 | |
Mouse | |
Protein G purified | |
Dendritic Cell Markers | |
9308 | |
Store at 4°C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction