Learn More
CD84 Rat anti-Mouse, PE, Clone: 1D3/CD84, BD Pharmingen™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566660
Catalog No. BDB566660
Add to cart
Description
Description
CD84 Monoclonal antibody specifically detects CD84 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD84
1D3/CD84
PE
Rat
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Mouse
Flow Cytometry
0.2 mg/mL
Conjugated, Purified
IgG1, κ
RUO
Primary
CD84