Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals CD84/SLAMF5 Antibody Pair [HRP]
Monoclonal Antibody Pair
Manufacturer: Novus Biologicals NBP2794395PLATES
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human CD84/SLAMF5.Specifications
156.25 pg/mL | |
Sandwich ELISA | |
RUO | |
This solid phase sandwich ELISA utilizes an antibody specific for Human CD84/SLAMF5. | |
CD84/SLAMF5 | |
5 Plates | |
Sandwich ELISA | |
CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B |
HRP | |
8832 | |
Antibody Pairs | |
156.25-10000 pg/mL | |
Storage is content dependent. | |
Mouse/Rabbit | |
Human | |
CD84 | |
1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 10000 pg/mL is recommended. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title