Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD84/SLAMF5 Mouse anti-Human, Clone: CD84.1.21, Novus Biologicals™

Mouse Monoclonal Antibody

$176.85 - $436.00

Specifications

Antigen CD84/SLAMF5
Clone CD84.1.21
Concentration 1.0 mg/mL
Dilution Flow Cytometry 1:100-1:200, Immunoprecipitation 1:10-1:500
Applications Flow Cytometry, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10065929
View Documents
Novus Biologicals
NB10065929
0.1 mg
Each for $436.00
Add to cart
 
NB017758
View Documents
Novus Biologicals
NB100659290.025MG
0.025 mg
Each for $176.85
Add to cart
 
Description

Description

CD84/SLAMF5 Monoclonal specifically detects CD84/SLAMF5 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.
Specifications

Specifications

CD84/SLAMF5
1.0 mg/mL
Flow Cytometry, Immunoprecipitation
Unconjugated
Mouse
Human
CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B
CD84
IgG1
Protein G purified
NB100-65929 recognizes the human CD84 cell surface antigen, a highly glycosylated cell surface antigen of 64-82kD. CD84 is predominantly expressed by B lymphocytes, monocytes and macrophages, although lower levels of expression have also been reported on T cells, granulocytes and platelets.
CD84.1.21
Flow Cytometry 1:100-1:200, Immunoprecipitation 1:10-1:500
Monoclonal
Purified
RUO
Q9UIB8
8832
Mouse pre-B cell line 300.19 transfected with CD84 cDNA.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit