Learn More
CD86 (B7-2) Mouse anti-Human, Baboon, Cynomolgus, Rhesus, BB700, Clone: 2331 (FUN-1), BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566474
Catalog No. BDB566474
Add to cart
Description
Description
CD86 (B7-2) Monoclonal antibody specifically detects CD86 (B7-2) in Human,Baboon,Cynomolgus,Rhesus samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD86 (B7-2)
2331 (FUN-1)
Conjugated, Purified
IgG1, κ
RUO
Primary
CD86 (B7-2)
Flow Cytometry
BB700
Mouse
25 Tests
Refrigerate (2 to 8°C)
Monoclonal
Human, Baboon, Cynomolgus, Rhesus Monkey