Learn More
CD86 Mouse anti-Baboon,Cynomolgus,Human,Rhesus, Brilliant Ultraviolet 737, Clone: 2331 (FUN-1), BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 612785
Catalog No. BDB612785
Add to cart
Description
Description
The 2331 (FUN-1) monoclonal antibody specifically recognizes a 75 kDa transmembrane cell surface protein, CD86 (B70/B7-2), expressed primarily on monocytes, dendritic cells and activated B cells. Competitive binding assays demonstrate that, while both 2331 (FUN-1) and IT2.2 (anti-CD86, Cat. No. 555663) antibodies specifically recognize the same molecule, they react with different epitopes. CD86 is the second ligand for CD28 and CTLA-4 and may play an important role in co-stimulation of T cells in primary immune response. The 2331 (FUN-1) antibody blocks the costimulatory activity of CD86 when tested in functional studies.
The antibody was conjugated to BD Horizon™ BUV737 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye is a tandem fluorochrome of BD Horizon BUV395 with an Ex Max of 348-nm and an acceptor dye with an Em Max at 737-nm. BD Horizon Brilliant BUV737 can be excited by the ultraviolet laser (355 nm) and detected with a 740/35 filter. Due to the excitation of the acceptor dye by other laser lines, there may be significant spillover into channels detecting Alexa Fluor™ 700-like dyes (e.g., 712/20-nm filter).
Specifications
Specifications
CD86
2331 (FUN-1)
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
IgG1, κ
25 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Ultraviolet 737
Affinity Purified
Human HBL-1 Cell Line
Affinity Chromatography
RUO
Primary
Baboon, Cynomolgus, Human, Rhesus Monkey