Learn More
CD86 Mouse anti-Human, BB515, Clone: 2331 (FUN-1), BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564545
Catalog No. BDB564545
Add to cart
Description
Description
CD86 Monoclonal antibody specifically detects CD86 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
Specifications
Specifications
CD86
2331 (FUN-1)
Vol. Per test: 5μL
Purified
Murine
Mouse BALB/c IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Monoclonal
Flow Cytometry
BB515
Aqueous buffered solution containing ≤0.09% sodium azide.
B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; BU63; CD28LG2; LAB72
Human HBL-1 Cell Line
25 Tests
Cell Biology
Primary
Baboon, Cynomolgus, Human, Rhesus
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.