Learn More
CD88 (C5aR1) Mouse anti-Human, PE, Clone: S5/1, eBioscience™ Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  LIFE TECHNOLOGIES 12088842
Encompass
Catalog No. 12-088-842
Add to cart
Description
Description
This S5/1 monoclonal antibody recognizes human CD88 also known as C5aR1. C5aR1 is a complement receptor widely expressed on cells of the myeloid lineage. Clone S5/1 has been reported to block C5a binding. Applications Reported: This S5/1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This S5/1 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount ( μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
CD88 (C5aR1) Monoclonal antibody specifically detects CD88 (C5aR1) in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
CD88 (C5aR1)
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P21730
Mouse
Affinity chromatography
RUO
728
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
S5/1
PE
C5AR1
C5A; C5a anaphylatoxin chemotactic receptor; C5a anaphylatoxin chemotactic receptor 1; C5a anaphylatoxin chemotactic receptor-like; C5a anaphylatoxin receptor; C5a ligand; C5a Receptor; C5ar; C5a-R; C5AR1; C5r1; CD88; CD88 antigen; complement C5a receptor; complement C5a receptor 1; complement component 5 receptor 1; complement component 5, receptor 1; complement component 5a receptor 1; D7Msu1; LOC100352978
BSA-Conjugated Peptide aa. 1-31
100 Tests
Primary
Human
Antibody
IgG2a κ