Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8a Mouse anti-Human, Clone: C8/144B, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 14008580

 View more versions of this product

Catalog No. 50-112-2631


Add to cart

Description

Description

Description: The monoclonal antibody C8/144B recognizes the human CD8a molecule, an approximately 32-34 kDa cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class 1 and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells. Applications Reported: This C8/144B antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This C8/144B antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tonsil tissue using IHC Antigen Retrieval Solution - High pH (cat. 00-4956). This can be used at less than or equal to 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.

CD8 (Cluster of Differentiation 8) is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediate efficient cell-cell interactions within the immune system. The CD8 antigen acts as a co-receptor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20-35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer (NK) cells, subpopulations of peripheral blood null cells, thymocytes and bone marrow cells. The CD8 co-receptor functions as either a homodimer composed of two alpha chains, or as a disulfide-linked heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. The majority of CD8+ T cells express CD8 as a alpha/beta heterodimer. In HIV, the HIV-2 envelope glycoprotein binds CD8 alpha chain (but not the beta chain).
Specifications

Specifications

CD8a
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P01732
CD8, Leu-2, MAL, p32, CD8a
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
C8/144B
Unconjugated
CD8A
Liquid
CD8A
IgG1, kappa
25 μg
4° C
Primary
925
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.