Learn More
CD8A Mouse anti-Human, FITC, Clone: COV8, Abnova™
Mouse monoclonal antibody raised against human CD8A.
Manufacturer:  Abnova Corporation MAB12379
Catalog No. 89-933-884
Add to cart
Description
Description
The CD8 antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen acts as a corepressor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The coreceptor functions as either a homodimer composed of two alpha chains, or as a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. This gene encodes the CD8 alpha chain isoforms. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
CD8A
COV8
Mouse monoclonal antibody raised against human CD8A.
In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA)
CD8/Leu2/MAL/p32
Mouse
IgG1
50 Reactions
Store at 4°C. Do not freeze.
Monoclonal
Human
Flow Cytometry, Immunofluorescence
FITC
Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user.
Liquid
CD8A
CD8A
Protein A/G affinity chromatography
RUO
Primary
925