Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CD8a, Mouse, FITC, Clone: RPA-T8,

Mouse Monoclonal Antibody

$51.50 - $441.50

Specifications

Antigen CD8a
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone RPA-T8
Host Species Mouse
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
MABF1673MI View Documents MilliporeSigma™
MABF1673
25 tests FITC Each for $51.50
Add to cart
 
MABF1674MI View Documents MilliporeSigma™
MABF1674
100 tests FITC Each for $104.00
Add to cart
 
MABF1675MI View Documents MilliporeSigma™
MABF1675
500 tests FITC Each for $441.50
Add to cart
 
Description

Description

The RPA-T8 antibody is specific for the 32-34kDa alpha chain of human CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor for antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development, and cytotoxic effector functions. CD8+ cytotoxic T cells (CTLs) play an important role in inducing cell death of tumor cells, as well as cells infected by virus, bacteria or parasites.The RPA-T8 antibody is widely used as a phenotypic marker for CD8 on cytotoxic T cells, thymocytes, as well as on certain cell types that do not also express the TCR, including some NK cells and lymphoid dendritic cells. It is cross-reactive with CD8 in several non-human species, including Baboon, Chimpanzee, Cynomolgus and Rhesus. If used together with an alternative Anti-Human CD8a clone, Hit8a, the RPA-T8 antibody will not block binding of Hit8a to CD8a.
Specifications

Specifications

CD8a
Monoclonal
Mouse
Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2.
Stable for 1 year at 2°-8°C from date of receipt. Note: It is recommended to store the product undiluted at 2°-8°C and protected from prolonged exposure to light. Do not freeze.
Primary
RPA-T8
Purified
Flow Cytometry
RUO

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit