Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8a Rabbit anti-Human, Unconjugated, Clone: 1B19, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen CD8a
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB101025MI
View Documents
MilliporeSigma
ZRB101025UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB10104XMI
View Documents
MilliporeSigma
ZRB10104X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

CD8a Monoclonal antibody specifically detects CD8a in Human samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

CD8a
Unconjugated
Rabbit
T-cell surface glycoprotein CD8 alpha chain;T-lymphocyte differentiation antigen T8/Leu-2;CD8a
His-tagged recombinant fragment corresponding to 143 amino acids from the extracellular domain of human CD8 alpha.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Purified
Human
CD8A, MAL
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit