Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 alpha Rat anti-Mouse, PE-Cyanine5.5, Clone: 5H10, Invitrogen™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MCD0818

Catalog No. 50-113-8095


Add to cart

Description

Description

CD8 (Cluster of Differentiation 8) is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediate efficient cell-cell interactions within the immune system. The CD8 antigen acts as a co-receptor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20-35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer (NK) cells, subpopulations of peripheral blood null cells, thymocytes and bone marrow cells. The CD8 co-receptor functions as either a homodimer composed of two alpha chains, or as a disulfide-linked heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. The majority of CD8+ T cells express CD8 as a alpha/beta heterodimer. In HIV, the HIV-2 envelope glycoprotein binds CD8 alpha chain (but not the beta chain).
Specifications

Specifications

CD8 alpha
Monoclonal
PE-Cyanine5.5
Cd8a
Liquid
Cd8a
Con A-stimulated BALB/c splenic T cells
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
5H10
PBS with BSA and 0.1% sodium azide
P01731
CD8, Leu-2, MAL, p32, CD8a
Rat
IgG2b
500 μL
4° C
Primary
12525
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit