Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8b Mouse Anti-Rat, Brilliant Violet 711, Clone: 341, BD Optibuild™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 742919

Catalog No. BDB742919


Add to cart

Description

Description

The 341 monoclonal antibody specifically recognizes theβchain of the CD8 antigen on most thymocytes and a subpopulation of mature T lymphocytes (ie, MHC class I-restricted T cells, including most T suppressor/cytotoxic cells). The CD8αandβchains (CD8a and CD8b, respectively) form a heterodimer on the surface of most thymocytes and thymus-dependent T suppressor/cytotoxic cells, whereas intestinal intraepithelial lymphocytes, many CD8+ T cells of athymic rats, many activated CD4+ T cells, and most NK cells express CD8a without CD8b. It has been suggested that the expression of the CD8a/CD8b heterodimer is restricted to thymus-derived T lymphocytes. CD8 is an antigen co-receptor on the T cell surface which interacts with MHC class I molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. Macrophages have also been reported to express CD8αandβchains, which are involved in signal transduction. The 341 mAb blocks proliferative and cytotoxic in vitro responses of CD8+ effectors to allogeneic cells.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
Specifications

Specifications

CD8b
341
Brilliant Violet 711
Cd8b, CD8beta, CD8 beta, CD8-beta, CD8β
CD8-positive Wistar rat splenic T-cell hybridomas
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
Affinity Purified
RUO
Mouse
Mouse BALB/c IgG1 κ
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit