Learn More
CD8b Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 212561AP
Catalog No. 50-173-3340
Add to cart
Description
Description
The CD8B antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen, acting as a coreceptor, and the T-cell receptor on the T lymphocyte recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The functional coreceptor is either a homodimer composed of two alpha chains, or a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. This gene encodes the CD8 beta chain isoforms. Multiple alternatively spliced transcript variants encoding distinct membrane associated or secreted isoforms have been described. A pseudogene, also located on chromosome 2, has been identified.
Specifications
Specifications
CD8b
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P10966
CD8, CD8b, CD8b molecule, CD8B1, Leu2, Ly3, LYT3
Rabbit
IgG
150 μL
-20°C
Primary
926
ELISA
Unconjugated
CD8B
Liquid
CD8B
CD8b Fusion Protein Ag15805
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human