Learn More
CD9 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0180-5H15, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212184AF594
Catalog No. NP212184A94
Add to cart
Description
Description
CD9 Monoclonal antibody specifically detects CD9 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD9
MM0180-5H15
Flow Cytometry
BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CD9
Human CD9
0.1 mL
Primary
928