Learn More
CD9 Mouse anti-Human, Clone: VJ1/20 (APC), Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13771
Catalog No. 89-033-836
Add to cart
Description
Description
Mouse monoclonal antibody raised against human CD9.
  • High specificity
  • High reactivity
Specifications
Specifications
CD9
VJ1/20
Mouse monoclonal antibody raised against human CD9.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
CD9
TNF activated HUVEC cells.
Protein A/G purification
RUO
Primary
928
Flow Cytometry
APC
Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user.
P21926
5H9|BA2|BTCC-1|DRAP-27|GIG2|MIC3|MRP-1|P24|TSPAN29
Mouse
IgG2a
100 Reactions
Store in the dark at 4°C. Avoid prolonged exposure to light.
Monoclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)