Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Mouse anti-Human, PE-Cyanine7, Clone: MM0180-5H15, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212184PECY7

Catalog No. N212184PEC7


Add to cart

Description

Description

CD9 Monoclonal antibody specifically detects CD9 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD9
MM0180-5H15
Flow Cytometry
Conjugated, Purified
CD9
Human CD9
Protein G purified
Cancer, Embryonic Stem Cell Markers, Stem Cell Markers
Primary
928.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit