Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Rat anti-Mouse, Clone: 4.1F12, MilliporeSigma™

Rat Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen CD9
Clone 4.1F12
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABT1560100
View Documents
MilliporeSigma
MABT1560100UL
100 μL
Each for $387.00
Add to cart
 
MABT156025
View Documents
MilliporeSigma
MABT156025UL
25 μL
Each for $153.40
Add to cart
 
Description

Description

CD9 Monoclonal antibody specifically detects CD9 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

CD9
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Purified
RUO
Purified rat monoclonal antibody IgG2c in PBS without azide.
Monoclonal
Protein G purified
4.1F12
Unconjugated
Rat
Mouse
Mouse platelets fused with X63 mouse myeloma.
Primary
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit