Learn More
CD9 Rat anti-Mouse, Janelia Fluor 646, Clone: EM-04, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP144876JF646
Catalog No. NBP44876646
Add to cart
Description
Description
CD9 Monoclonal antibody specifically detects CD9 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
CD9
EM-04
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Purified
CD9
Permeabilized murine bone marrow-derived mast cells (BMMC).
Protein A purified
Cancer, Embryonic Stem Cell Markers, Stem Cell Markers
Primary
928
Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Rat
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
The rat monoclonal antibody EM-04 recognizes CD9 antigen, a 24 kDa transmembrane protein expressed on platelets, monocytes, pre-B lymphocytes, granulocytes and activated T lymphocytes.