Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90.2, Rat anti-Mouse, Biotin, Clone: 53-2.1, Novus Biologicals™

Rat Monoclonal Antibody

$366.50 - $450.50

Specifications

Antigen CD90.2
Clone 53-2.1
Applications Flow Cytometry
Host Species Rat
Target Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB004395
View Documents
Novus Biologicals
NBP143609100UG
100 ug
Each for $366.50
Add to cart
 
NB004396
View Documents
Novus Biologicals
NBP143609500UG
500 ug
Each for $450.50
Add to cart
 
Description

Description

CD90.2 Monoclonal antibody specifically detects CD90.2 in Mouse Samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD90.2
Flow Cytometry
Mouse
Primary
53-2.1
Rat
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit