Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Supplier: LIFE TECHNOLOGIES M028T03B06

This item is not returnable.
View return policy
Description
CD90.2 (Thy-1.2) Monoclonal antibody specifically detects CD90.2 (Thy-1.2) in Mouse samples. It is validated for Flow CytometrySpecifications
CD90.2 (Thy-1.2) | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P01831 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
53-2.1 | |
NovaFluor Blue 610-70S | |
Thy1 | |
CD90; FLJ33325; T25; Thy 1.2; Thy1; Thy-1; Thy-1 antigen; thy-1 membrane glycoprotein; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta | |
Affinity chromatography | |
RUO | |
21838 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title