Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Specifications

Antigen CD90.2 (Thy-1.2)
Clone 53-2.1
Concentration 0.1 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
M028T02B06
Encompass
View Documents
LIFE TECHNOLOGIES
M028T02B06
25 μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
M028T03B06
Encompass
View Documents
LIFE TECHNOLOGIES
M028T03B06
100 μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

CD90.2 (Thy-1.2) Monoclonal antibody specifically detects CD90.2 (Thy-1.2) in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

CD90.2 (Thy-1.2)
0.1 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
CD90; FLJ33325; T25; Thy 1.2; Thy1; Thy-1; Thy-1 antigen; thy-1 membrane glycoprotein; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta
IgG2a κ
Affinity chromatography
Antibody
53-2.1
Flow Cytometry
NovaFluor Blue 610-70S
Rat
Mouse
P01831
21838
Primary
4° C, store in dark, DO NOT FREEZE!
Thy1
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit