Learn More
CD90/Thy1 Mouse anti-Human, Alexa Fluor™ 405, Clone: 783922, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB20671V100UG
Catalog No. FB20671V100
Add to cart
This item is not returnable. View return policy
Description
Description
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD90/Thy1
783922
Flow Cytometry 0.25-1 μg/106 cells
P04216
Mouse
IgG2a
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human CD90/THY1 in direct ELISAs.
Flow Cytometry
Alexa Fluor™ 405
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide
CD90, CD90 antigen, CDw90, FLJ33325, Thy1, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Chinese Hamster Ovary cell line, CHO-derived human CD90/THY1, Met1-Ser141, Accession No. P04216
100 μg
Primary
7070
Human