Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90 Mouse anti-Human, FITC, Clone: 5E10, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen A15761

Catalog No. 50-113-7095


Add to cart

Description

Description

CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. It is expressed in murine T cells, thymocytes, neural cells, Kupffer ft.s cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain.
Specifications

Specifications

CD90
Monoclonal
FITC
THY1
Liquid
THY1
Human CD90
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
5E10
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P04216
THY1, Thy-1 cell surface antigen, FLJ33325, Thy-1 T-cell antigen, Thy-1 membrane glycoprotein
Mouse
IgG1
100 tests
4° C
Primary
7070
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit