Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD90/Thy1 Mouse anti-Human, Primate, Allophycocyanin/Cy7, Clone: F15-42-1, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP233316APCCY7
Description
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human, Primate samples. It is validated for Flow Cytometry.Specifications
CD90/Thy1 | |
F15-42-1 | |
Flow Cytometry | |
Conjugated, Purified | |
THY1 | |
Purified human brain Thy1. | |
Protein G purified | |
Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
Primary | |
7070.0 | |
Human, Primate |
Flow Cytometry | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
CD90 (F15-42-1) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title