Learn More
CD94 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0181-5F26, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212185AF594
Catalog No. NP21218A94
Add to cart
Description
Description
CD94 Monoclonal antibody specifically detects CD94 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD94
MM0181-5F26
Flow Cytometry
CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
KLRD1
Human CD94
0.1 mL
Primary
3824