Learn More
CD94 Mouse anti-Human, BB700, Clone: HP-3D9, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566534
 View more versions of this product
Catalog No. BDB566534
Add to cart
Description
Description
CD94 Monoclonal antibody specifically detects CD94 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD94
HP-3D9
Purified
IgG1, κ
RUO
Primary
Human
Flow Cytometry
BB700
Mouse
100 Tests
Refrigerate (2 to 8°C)
Monoclonal