Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD94 Mouse anti-Human, BB700, Clone: HP-3D9, BD Horizon™

Mouse Monoclonal Antibody

$175.00 - $330.00

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen CD94
Conjugate BB700
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566535
View Documents
BD Biosciences
566535
25 Tests
Each for $175.00
Add to cart
 
BDB566534
View Documents
BD Biosciences
566534
100 Tests
Each for $330.00
Add to cart
 
Description

Description

CD94 Monoclonal antibody specifically detects CD94 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Monoclonal
Primary
CD94
BB700
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit