Learn More
CD94 Mouse, anti-Human, R718, Clone: HP-3D9, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 752211
Catalog No. BDB752211
Add to cart
Description
Description
The HP-3D9 monoclonal antibody specifically binds to CD94 which is also known as KP43. CD94 is expressed on the cell surface as a ~70 kDa, disulfide-linked, type II transmembrane glycoprotein dimer. It is encoded by KLRD1 (Killer cell lectin like receptor D1) which belongs to the C-type lectin superfamily. CD94 is expressed on natural killer (NK) cells, especially activated NK cells. It is also expressed on γ/δ TCR+ T lymphocytes, NK-T cells, and on some CD8+CD56+ α/β TCR+ cells. CD94 associates with various NKG2 receptors to form receptors for HLA class I molecules and plays a role in regulating cellular adhesion and activation. The HP-3D9 antibody can reportedly inhibit the cytolytic activity of activated NK cells.
Specifications
Specifications
CD94
HP-3D9
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Human