Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD95 (Fas) Mouse anti-Human, Baboon, Cynomolgus, Rhesus, BB700, Clone: DX2, BD Horizon™
Mouse Monoclonal Antibody
$172.00 - $406.00
Specifications
Antigen | CD95 (Fas) |
---|---|
Clone | DX2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Blue 700 (BB700) |
Description
CD95 (Fas) Monoclonal antibody specifically detects CD95 (Fas) in Human,Baboon,Cynomolgus,Rhesus samples. It is validated for Flow Cytometry.Specifications
CD95 (Fas) | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |
DX2 | |
Monoclonal | |
Mouse | |
Human, Baboon, Cynomolgus, Rhesus Monkey | |
Primary | |
CD95 (Fas) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title