Learn More
CD96 Mouse anti-Human, Alexa Fluor 405, Clone: 628211, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB6199V100UG
Catalog No. FAB6199V100
Add to cart
This item is not returnable. View return policy
Description
Description
CD96 Monoclonal specifically detects CD96 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD96
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
NP_005807
CD96
Mouse myeloma cell line NS0-derived recombinant human CD96 Val22-Asp501 Accession # NP_005807
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human CD96 in direct ELISAs.
Flow Cytometry
628211
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD96, CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILE, TACTILEincreased late expression, T-cell surface protein tactile
Mouse
IgG1
RUO
Primary
10225
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)