Learn More
CD96 Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0690-1H29, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP236784AF750
Catalog No. NP236784A70
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD96 Monoclonal antibody specifically detects CD96 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CD96
MM0690-1H29
Western Blot
CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILEincreased late expression, T-cell surface protein tactile
Mouse
IgG2
Store at 4°C in the dark.
Monoclonal
This antibody recognizes human CD96 V2 isoform by direct ELISA and WB.
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CD96
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Recombinant human CD96 V2 isoform.
0.1 mL
Primary
10225
Human