Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD96 Mouse anti-Human, Biotin, Clone: MM0690-1H29, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236784B

Catalog No. NBP236784B


Add to cart

Description

Description

CD96 Monoclonal antibody specifically detects CD96 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CD96
MM0690-1H29
Western Blot
Purified
CD96
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Recombinant human CD96 V2 isoform.
Protein G purified
Store at 4C in the dark.
Monoclonal
This antibody recognizes human CD96 V2 isoform by direct ELISA and WB.
Western Blot
Biotin
PBS with 0.05% Sodium Azide
CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILEincreased late expression, T-cell surface protein tactile
Mouse
IgG2
0.1 ml
Primary
10225
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit