Learn More
CD96 Mouse anti-Human, DyLight 650, Clone: MM0690-1H29, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236784C
Catalog No. NBP236784C
Add to cart
Description
Description
CD96 Monoclonal antibody specifically detects CD96 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CD96
MM0690-1H29
Western Blot
Purified
CD96
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Recombinant human CD96 V2 isoform.
Protein G purified
Store at 4C in the dark.
Monoclonal
This antibody recognizes human CD96 V2 isoform by direct ELISA and WB.
Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILEincreased late expression, T-cell surface protein tactile
Mouse
IgG2
0.1 ml
Primary
10225
Human