Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD97 Rabbit anti-Human, DyLight 650, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP299764C
Description
CD97 Polyclonal antibody specifically detects CD97 in Human samples. It is validated for Western Blot, ELISASpecifications
CD97 | |
DyLight 650 | |
50mM Sodium Borate | |
CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
Produced in rabbits immunized with purified, recombinant CD97 (NP_001775.2; Met 1-Gln 398). | |
Affinity purified | |
RUO | |
Store at 4°C in the dark. | |
Polyclonal | |
Human |
ELISA, Western Blot | |
Western Blot, ELISA | |
Purified | |
Rabbit | |
IgG | |
0.1 mL | |
GPCR, Immunology | |
Primary | |
976 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title