Learn More
CD97 Mouse anti-Human, FITC, Clone: MEM-180, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15738
Catalog No. 50-113-7087
Add to cart
Description
Description
CD97 is a 75-85kDa monomeric glycoprotein that is present on the surface of most activated leucocytes. It is a receptor involved in both cell adhesion and signaling. CD97 is a member of the G protein coupled receptor family. It is expressed abundantly in cells of hematopoietic origin and is markedly upregulated on activated T and B cells. ESTs have been isolated from a wide variety of tissue libraries.
Specifications
Specifications
CD97
Monoclonal
FITC
ADGRE5
Liquid
ADGRE5
Human CD97
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MEM-180
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P48960
CD97, CD97 molecule, Leukocyte antigen CD97, TM7LN1, CD97 antigen
Mouse
IgG1
100 tests
4° C
Primary
976