Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD97 Rabbit anti-Human, DyLight 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP299764UV

Catalog No. NB062781


Add to cart

Description

Description

CD97 Polyclonal antibody specifically detects CD97 in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

CD97
DyLight 350
50mM Sodium Borate
CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1
Produced in rabbits immunized with purified, recombinant CD97 (NP_001775.2; Met 1-Gln 398).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
ELISA, Western Blot
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
GPCR, Immunology
Primary
976
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit